missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPTX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20656-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to NPTX1.
NPTX1 is a member of the neuronal pentraxin family. Human neuronal pentraxin 1 is similar to rat NP1 which is a binding protein for the snake venom toxin taipoxin.Spécification
NPTX1 | |
Polyclonal | |
Unconjugated | |
NPTX1 | |
Neuronal pentraxin 1, neuronal pentraxin I, NPTX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18164, 266777, 4884 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P47971, Q15818, Q62443 | |
Nptx1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |