Learn More
NeutraKine™VEGF165/121 Mouse anti-Human, Clone: 6C3D5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 69025-1-IG-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NeutraKine®VEGF165/121 Monoclonal antibody specifically detects NeutraKine®VEGF165/121 in Human samples. It is validated for Neutralization, ELISASpécification
NeutraKine™VEGF165/121 | |
Monoclonal | |
Unconjugated | |
BC010129 | |
Protein G purification | |
Primary | |
Human | |
Antibody | |
IgG1 |
Neutralization, ELISA | |
6C3D5 | |
Sterile PBS. | |
Mouse | |
100 μg | |
7422 | |
Store at -20°C | |
Liquid |