Learn More
NeutraKine™ PDGF-BB Mouse anti-Human, Unconjugated, Clone: 4G11C7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 69020-1-IG-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The NeutraKine™ PDGF-BB antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human NeutraKine™ PDGF-BB. This antibody recognizes human antigen. The NeutraKine™ PDGF-BB antibody has been validated for the following applications: Neutralization, ELISA analysis.
Spécification
NeutraKine™ PDGF-BB | |
Monoclonal | |
Unconjugated | |
100 μg | |
5155 | |
IgG1 |
Neutralization | |
4G11C7 | |
Mouse | |
Primary | |
Human |