missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20140-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to MYH4.
Muscle contraction. Weiss A., J. Mol. Biol. 290:61-75(1999). Denis N. J., Proteomics 7:868-874(2007).Spécification
MYH4 | |
Polyclonal | |
Unconjugated | |
MYH4 | |
MYH2B, MYH4, MyHC 2B, MyHC IIb, Myosin 4, Myosin heavy chain 2b, Myosin heavy chain 4, Myosin heavy chain IIb | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17884, 4622 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5SX39, Q9Y623 | |
Myh4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |