missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLF1IP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13186-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The function of this protein remains unknown.Spécification
MLF1IP | |
Polyclonal | |
Unconjugated | |
CENPU | |
CENP 50, CENP U, CENP U(50), CENP50, CENPU, centromere protein of 50 kDa, Centromere protein U, ICEN24, KLIP1, MLF1 interacting protein, MLF1IP, PBIP1, Polo box interacting protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
306464, 71876, 79682 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4V8G7, Q71F23, Q8C4M7 | |
Cenpu | |
MLF1IP Fusion Protein Ag3858 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |