missing translation for 'onlineSavingsMsg'
Learn More

MAX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Spécification

Antigène MAX
Concentration 0.37 mg/mL
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16806813
Afficher les documents Détails de la promotion
Proteintech
10426-1-AP-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16896803
Afficher les documents Détails de la promotion
Proteintech
10426-1-AP-150UL
150 μL
410.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Bands recognized by this antibody are much larger than predicted.

The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include Mad, Mxi1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature for some of them is unknown.
Spécification

Spécification

MAX
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Human, Rat
P28574, P52164, P61244
17187, 4149, 60661
MAX Fusion Protein Ag0680
Primary
-20°C
MAX
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8, MAX, MYC associated factor X, orf1, Protein max
Max
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications