missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10426-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Bands recognized by this antibody are much larger than predicted.
The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include Mad, Mxi1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature for some of them is unknown.Spécification
MAX | |
Polyclonal | |
Unconjugated | |
MAX | |
bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8, MAX, MYC associated factor X, orf1, Protein max | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17187, 4149, 60661 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P28574, P52164, P61244 | |
Max | |
MAX Fusion Protein Ag0680 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |