Learn More
mAID tag Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28209-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The mAID tag antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of arabidopsis thaliana mAID tag. This antibody recognizes human antigen. The mAID tag antibody has been validated for the following applications: ELISA analysis.
Spécification
mAID tag | |
Polyclonal | |
Unconjugated | |
20 μL | |
Human |
ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
IgG |