Learn More
mAID tag Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28209-1-AP-150UL
Code produit. 16836055
  • 399.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
The mAID tag antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of arabidopsis thaliana mAID tag. This antibody recognizes human antigen. The mAID tag antibody has been validated for the following applications: ELISA analysis.
Spécification
Spécification
mAID tag
Polyclonal
Unconjugated
150 μL
Human
ELISA
300 μg/mL