missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1191 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20637-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
KIAA1191 | |
Polyclonal | |
Unconjugated | |
KIAA1191 | |
KIAA1191, p60MONOX, UPF0498 protein KIAA1191 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57179, 97820 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96A73 | |
4833439L19Rik, KIAA1191 | |
KIAA1191 Fusion Protein Ag14694 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |