missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KATNAL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20166-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.Spécification
KATNAL1 | |
Polyclonal | |
Unconjugated | |
KATNAL1 | |
katanin p60 subunit A like 1, KATNAL1, p60 katanin like 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84056 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BW62 | |
KATNAL1 | |
KATNAL1 Fusion Protein Ag13960 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |