Learn More
KATNAL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20166-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16809674
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.
Spécification
Spécification
KATNAL1
Polyclonal
Unconjugated
KATNAL1
katanin p60 subunit A like 1, KATNAL1, p60 katanin like 1
Rabbit
Antigen Affinity Chromatography
RUO
84056
-20°C
Liquid
Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BW62
KATNAL1
KATNAL1 Fusion Protein Ag13960