missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANSL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27261-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
KANSL2 | |
Polyclonal | |
Unconjugated | |
KANSL2 | |
C12orf41, KANSL2, KAT8 regulatory NSL complex subunit 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54934, 69612 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BQR4, Q9H9L4 | |
KANSL2 | |
KANSL2 Fusion Protein Ag25983 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |