missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jph2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51054-2-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibililty to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.Spécification
Jph2 | |
Polyclonal | |
Unconjugated | |
JPH2 | |
1110002E14Rik, JP 2, Jp2, Jph2, junctophilin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57158 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BR39 | |
JPH2 | |
Jph2 Fusion Protein Ag0468 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |