missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14069-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
INTS5 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.Spécification
INTS5 | |
Polyclonal | |
Unconjugated | |
INTS5 | |
INT5, integrator complex subunit 5, INTS5, KIAA1698 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109077, 309200, 80789 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P9B9, Q8CHT3 | |
INTS5 | |
INTS5 Fusion Protein Ag5129 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |