missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | IGF1 |
---|---|
Concentration | 333 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugué | Unconjugated |
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
16826185
|
Proteintech
28530-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16816185
|
Proteintech
28530-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
The IGF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IGF1. This antibody recognizes human antigen. The IGF1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
IGF1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG |
333 μg/mL | |
Polyclonal | |
Rabbit | |
3479 | |
Primary |