Learn More
IGF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28530-1-AP-20UL
Code produit. 16826185
  • 140.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
The IGF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IGF1. This antibody recognizes human antigen. The IGF1 antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
Spécification
IGF1
Polyclonal
Unconjugated
20 μL
3479
IgG
Western Blot, Immunohistochemistry
333 μg/mL