Learn More
HOXD9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20560-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The HOXD9 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human HOXD9. This antibody recognizes human antigen. The HOXD9 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Spécification
HOXD9 | |
Polyclonal | |
Unconjugated | |
20 μL | |
3235 | |
IgG |
Western Blot, Immunofluorescence, ELISA | |
167 μg/mL | |
Rabbit | |
Primary | |
Human |