Learn More
HOXD9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20560-1-AP-150UL
Code produit. 16860115
  • 399.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
The HOXD9 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human HOXD9. This antibody recognizes human antigen. The HOXD9 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Spécification
Spécification
HOXD9
Polyclonal
Unconjugated
150 μL
3235
IgG
Western Blot, Immunofluorescence, ELISA
167 μg/mL