missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55050-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to GSX1.
GSX1 gene ontology annotations related to this gene include adenohypophysis development; hypothalamus development; neuron fate commitment; spinal cord association neuron differentiation.Spécification
GSX1 | |
Polyclonal | |
Unconjugated | |
GSX1 | |
GS homeobox 1, Gsh 1, GSH1, GSX1, Homeobox protein GSH 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14842, 219409, 288457 | |
-20°C | |
Liquid |
Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31315, Q9H4S2 | |
Gsx1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |