Learn More
GLT25D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16768-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
GLT25D1 | |
Polyclonal | |
Unconjugated | |
COLGALT1 | |
GLT25D1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234407, 290637, 79709 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K297, Q8NBJ5 | |
Colgalt1 | |
GLT25D1 Fusion Protein Ag10346 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |