missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spezifikation
Antigène | FOXC1 |
---|---|
Concentration | 0.74 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Produktcode | Marke | Quantité | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantité | Preis | Menge & Verfügbarkeit | |||||
16836835
|
Proteintech
55365-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16826835
|
Proteintech
55365-1-AP-150UL |
150 μL |
399.00€
150µL |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Beschreibung
This antibody is specific to FOXC1. Phosphorylation modification of Foxc1 protein may be responsible for the larger molecular weight of detection compared with theoretical molecular weight.
FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintece of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.Spezifikation
FOXC1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q12948, Q61572 | |
17300, 2296 | |
Peptide | |
Primary | |
-20°C | |
FOXC1 |
0.74 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARA, FKHL7, forkhead box C1, Forkhead box protein C1, Forkhead related protein FKHL7, FOXC1, FREAC 3, FREAC3, IGDA, IHG1, IRID1, RIEG3 | |
Foxc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |