missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55365-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is specific to FOXC1. Phosphorylation modification of Foxc1 protein may be responsible for the larger molecular weight of detection compared with theoretical molecular weight.
FOXC1 is a protein belonging to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this protein is not known, however, it plays a role in the regulation of FGF19-FGFR4-MAPK pathway to promote both the development and maintece of anterior segment structures within the eye. Mutations in this gene cause various glaucoma, iridogoniodysgenesis anomaly, Peters anomaly that includes central corneal leukoma, absence of the posterior corneal stroma and Descemetmembrane and Axenfeld-Rieger anomaly characterized by posterior corneal embryotoxon, iris adhesion to the Schwalbe line, hypertelorism, hypoplasia of the malar bones, congenital absence of some teeth and mental retardation.Spécification
FOXC1 | |
Polyclonal | |
Unconjugated | |
FOXC1 | |
ARA, FKHL7, forkhead box C1, Forkhead box protein C1, Forkhead related protein FKHL7, FOXC1, FREAC 3, FREAC3, IGDA, IHG1, IRID1, RIEG3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17300, 2296 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12948, Q61572 | |
Foxc1 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |