Learn More
FMNL1 Mouse anti-Human, Unconjugated, Clone: 1B12A4, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67587-1-IG-20UL
Code produit. 16800306
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
The FMNL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FMNL1. This antibody recognizes human antigen. The FMNL1 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
FMNL1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1B12A4
Unconjugated
20 μL
752
IgG2b