Learn More
FMNL1 Mouse anti-Human, Unconjugated, Clone: 1B12A4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67587-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The FMNL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FMNL1. This antibody recognizes human antigen. The FMNL1 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
FMNL1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1B12A4 | |
Unconjugated | |
20 μL | |
752 | |
IgG2b |