Learn More
EPOR Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 30766-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The EPOR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human EPOR. This antibody recognizes human, mouse antigen. The EPOR antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
EPOR | |
450 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse | |
150 kDa | |
IgG |
Western Blot, ELISA | |
EPO R, EPOR, erythropoietin receptor | |
20 μL | |
2057 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |