missing translation for 'onlineSavingsMsg'
Learn More

Elastin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Spécification

Antigène Elastin
Concentration 0.16 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16854354
Afficher les documents
Proteintech
15257-1-AP-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16844354
Afficher les documents
Proteintech
15257-1-AP-150UL
150 μL
410.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

Elastin
Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
P15502, P54320
13717, 2006
Elastin Fusion Protein Ag7179
Primary
-20°C
ELN
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
elastin, ELN, SVAS, Tropoelastin, WBS, WS
ELN
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications