missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15257-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
Elastin | |
Polyclonal | |
Unconjugated | |
ELN | |
elastin, ELN, SVAS, Tropoelastin, WBS, WS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13717, 2006 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15502, P54320 | |
ELN | |
Elastin Fusion Protein Ag7179 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |