missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DSS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13639-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The product of this gene has been localized within the split hand/split foot malformation locus SHFM1 at chromosome 7. It has been proposed to be a candidate gene for the autosomal domit form of the heterogeneous limb developmental disorder split hand/split foot malformation type 1. In addition, it has been shown to directly interact with BRCA2. It also may play a role in the completion of the cell cycle.Spécification
DSS1 | |
Polyclonal | |
Unconjugated | |
SEM1 | |
DSS1, ECD, SEM1, SHFD1, Shfdg1, SHFM1, SHSF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7979 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60896 | |
SEM1 | |
DSS1 Fusion Protein Ag4573 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |