Learn More
DDX18 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28502-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The DDX18 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DDX18. This antibody recognizes human, mouse antigen. The DDX18 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
DDX18 | |
Polyclonal | |
Unconjugated | |
20 μL | |
8886 | |
IgG |
Western Blot, ELISA | |
400 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |