Learn More
CYP11A1 Mouse anti-Human, Unconjugated, Clone: 1C5D5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67264-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CYP11A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP11A1. This antibody recognizes human, mouse, rat antigen. The CYP11A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
CYP11A1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry | |
1C5D5 | |
Unconjugated | |
20 μL | |
1583 | |
IgG1 |