Learn More
CYP11A1 Mouse anti-Human, Unconjugated, Clone: 1C5D5, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67264-1-IG-150UL
Code produit. 16819795
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
The CYP11A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP11A1. This antibody recognizes human, mouse, rat antigen. The CYP11A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
CYP11A1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry
1C5D5
Unconjugated
150 μL
1583
IgG1