Learn More
COL4A3BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène COL4A3BP
Concentration 0.15 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications
Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16854264
Afficher les documents
Proteintech
15191-1-AP-20UL
20 μL
145.00€
20µL
16844264
Afficher les documents
Proteintech
15191-1-AP-150UL
150 μL
410.00€
150µL
Description
Description
CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.
Spécification
Spécification
COL4A3BP
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Rat, Human, Mouse
Q9EQG9, Q9Y5P4
10087, 365652, 68018
COL4A3BP Fusion Protein Ag7277
Primary
-20°C
COL4A3BP
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Ceramide transfer protein, CERT, CERTL, COL4A3BP, GPBP, hCERT, STARD11
Col4a3bp
IgG
Antigen Affinity Chromatography