missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL4A3BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15191-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.Spécification
COL4A3BP | |
Polyclonal | |
Unconjugated | |
COL4A3BP | |
Ceramide transfer protein, CERT, CERTL, COL4A3BP, GPBP, hCERT, STARD11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10087, 365652, 68018 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EQG9, Q9Y5P4 | |
Col4a3bp | |
COL4A3BP Fusion Protein Ag7277 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |