missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLMP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16127-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
May be involved in the cell-cell adhesion. May play a role in adipocyte differentiation and development of obesity. Is required for normal small intestine development.Spécification
CLMP | |
Polyclonal | |
Unconjugated | |
CLMP | |
ACAM, adipocyte adhesion molecule, ASAM, CLMP, CXADR-like membrane protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
286939, 71566, 79827 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K1G0, Q8R373, Q9H6B4 | |
Clmp | |
CLMP Fusion Protein Ag9111 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |