missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39/ENTPD1 Mouse anti-Human, Clone: 2D2H6, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60323-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
60323-1-Ig can recognize the soluble form of CD39 (38-478 aa) molecule.
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.Spécification
CD39/ENTPD1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49961 | |
ENTPD1 | |
CD39/ENTPD1 Fusion Protein Ag6165 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
2D2H6 | |
Unconjugated | |
ENTPD1 | |
ATPDase, CD39, Ecto apyrase, Ecto ATP diphosphohydrolase 1, Ecto ATPase 1, Ecto ATPDase 1, ENTPD1, NTPDase 1 | |
Mouse | |
Protein A | |
RUO | |
953 | |
-20°C | |
Liquid |