Learn More
CD22 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21894-1-AP-150UL
This item is not returnable.
View return policy
Description
The CD22 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CD22. This antibody recognizes human antigen. The CD22 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CD22 | |
Polyclonal | |
Unconjugated | |
150 μL | |
933 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human |