Learn More
CCT5 Mouse anti-Human, Unconjugated, Clone: 1B12E10, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67400-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CCT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT5. This antibody recognizes human, mouse, rat antigen. The CCT5 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
CCT5 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1B12E10 | |
Unconjugated | |
20 μL | |
22948 | |
IgG1 |