Learn More
CCR5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 17476-1-AP-20UL
Code produit. 16897364
  • 140.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
The CCR5 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CCR5. This antibody recognizes human antigen. The CCR5 antibody has been validated for the following applications: FC, IHC, ELISA analysis.
Spécification
Spécification
CCR5
Polyclonal
Unconjugated
20 μL
1234
IgG
Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
387 μg/mL