missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC115 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20636-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
May be involved in Golgi homeostasis.Spécification
CCDC115 | |
Polyclonal | |
Unconjugated | |
CCDC115 | |
CCDC115, ccp1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363213, 69668, 84317 | |
-20°C | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VE99, Q96NT0 | |
Ccdc115 | |
CCDC115 Fusion Protein Ag14677 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |