Learn More
CaMKII delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15443-1-AP-20UL
 Afficher plus de versions de ce produit
Code produit. 16814624
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement »
Découvrez plus d'offres spéciales
Description
Description
This antibody is specific to the C-term of delta chain, CaMKII delta. It can recognize most isoforms the gene encodes, from 50-72 kDa. It can recognize isoforms 2, 3, 4, 9, 11.
CaMKII delta belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns
Spécification
Spécification
CaMKII delta
Polyclonal
Unconjugated
CAMK2D
CaM kinase II subunit delta, CaMK II subunit delta, CAMK2D, CAMKD, CaMKII delta
Rabbit
Antigen Affinity Chromatography
RUO
108058, 24246, 817
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13557
CAMK2D
CaMKII delta Fusion Protein Ag7323
20 μL
Primary
Mouse, Human, Rat