missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17275-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
May be similar to that of authentic calmodulin and may actually compete with calmodulin by binding, with different affinities, to cellular substrates.Spécification
CALML3 | |
Polyclonal | |
Unconjugated | |
CALML3 | |
CALML3, calmodulin like 3, Calmodulin like protein 3, CaM like protein, CLP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
810 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P27482 | |
CALML3 | |
CALML3 Fusion Protein Ag11037 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |