missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène | CA3 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16854274
|
Proteintech
15197-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16844274
|
Proteintech
15197-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Carbonic anhydrase III (CAIII) is a member of carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal.Spécification
CA3 | |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P07451, P14141, P16015 | |
12350, 54232, 761 | |
CA3 Fusion Protein Ag7344 | |
Primary | |
-20°C | |
CA3 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CA III, CA3, CAIII, Car3, Carbonate dehydratase III, Carbonic anhydrase 3, Carbonic anhydrase III | |
Ca3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |