missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15197-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Carbonic anhydrase III (CAIII) is a member of carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal.Spécification
CA3 | |
Polyclonal | |
Unconjugated | |
CA3 | |
CA III, CA3, CAIII, Car3, Carbonate dehydratase III, Carbonic anhydrase 3, Carbonic anhydrase III | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12350, 54232, 761 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07451, P14141, P16015 | |
Ca3 | |
CA3 Fusion Protein Ag7344 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |