Learn More
c-MYC Mouse anti-Human, Unconjugated, Clone: 3D9C12, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67447-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The c-MYC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human c-MYC. This antibody recognizes human, mouse, rat antigen. The c-MYC antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Spécification
c-MYC | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry, ELISA | |
3D9C12 | |
Unconjugated | |
20 μL | |
4609 | |
IgG1 |