Learn More
Berkshire™ 360 Isolator Cleaning System
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Marque: Berkshire™ EC360.NW.40
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
EasyClean 360 ICT Nonwoven Elastic Covers (17-987-363)
- Made from Berkshire’s Durx™ 670 55% cellulose/45% polyester nonwoven material
- Absorbent rayon inner layer
- Elastic edge allows easy installation and secure fit on mop head
- Recommended for use in ISO Class 5-8/EU Grade C-D environments
Spécification
Nonwoven Cellulose/Polyester Elastic Covers | |
2 Ply | |
EasyClean™ 360 Isolator Cleaning Tool | |
Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments. | |
55% cellulose/45%polyester nonwoven elastic |
Autoclavable | |
5 nonwoven elastic covers/pack | |
Mop Covers | |
ISO Class 5-8/EU Grade C-D |