Learn More
Berkshire™ 360 Isolator Cleaning System
Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Marque:  Berkshire™ EC360.NW.40
Code produit. 15438500
  • 785.00€ / 10 boîtes
Expédition estimée: 11-12-2024
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
EasyClean 360 ICT Nonwoven Elastic Covers (17-987-363)
  • Made from Berkshire’s Durx™ 670 55% cellulose/45% polyester nonwoven material 
  • Absorbent rayon inner layer
  • Elastic edge allows easy installation and secure fit on mop head
  • Recommended for use in ISO Class 5-8/EU Grade C-D environments
Spécification
Spécification
Nonwoven Cellulose/Polyester Elastic Covers
2 Ply
EasyClean™ 360 Isolator Cleaning Tool
Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
55% cellulose/45%polyester nonwoven elastic
Autoclavable
5 nonwoven elastic covers/pack
Mop Covers
ISO Class 5-8/EU Grade C-D