Learn More
BAIAP2L2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16738-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The BAIAP2L2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAIAP2L2. This antibody recognizes human antigen. The BAIAP2L2 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
BAIAP2L2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
80115 | |
IgG |
Western Blot, ELISA | |
253 μg/mL | |
Rabbit | |
Primary | |
Human |