Learn More
BAIAP2L2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16738-1-AP-150UL
Code produit. 16866464
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
The BAIAP2L2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAIAP2L2. This antibody recognizes human antigen. The BAIAP2L2 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
BAIAP2L2
Polyclonal
Unconjugated
150 μL
80115
IgG
Western Blot, ELISA
253 μg/mL