missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF7IP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14699-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ATF7IP is a multifunctional nuclear protein that associates with heterochromatin. It can act as a transcriptional coactivator or corepressor depending upon its binding partners.Spécification
ATF7IP | |
Polyclonal | |
Unconjugated | |
ATF7IP | |
ATF7IP, MCAF, p621 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
312800, 54343, 55729 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6VMQ6, Q7TT18 | |
Atf7ip | |
ATF7IP Fusion Protein Ag6341 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |