missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10312-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ADP-ribosylation factor-like 4A is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4A is similar to ARL4C and ARL4D and each has a nuclear localization signal and an unusually high guaninine nucleotide exchange rate. ARL4A is located in both the nuclear and extranuclear cell compartments. Multiple transcript variants encoding the same protein have been found for this gene.Spécification
ARL4 | |
Polyclonal | |
Unconjugated | |
ARL4A | |
ARL4, ARL4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10124, 11861, 29308 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P40617, P61213, P61214 | |
ARL4A | |
ARL4 Fusion Protein Ag0342 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |